Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 5-Bromo-2-morpholin-1-yl-pyrimidine

Catalog No. SYX002771G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
SYX002771G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. SYX002771G Supplier Sigma Aldrich Supplier No. SYX002771G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • Phase behavior and phase separation kinetics of ternary nonionic microemulsion system composed of pentaethylene glycol dodecyl ether, water and 1-chlorotetradecane was studied.1-chlorotetradecane has been used:
  • In preparation of nonionic microemulsion of pentaoxyethylene glycol dodecyl ether, water and 1-chlorotetradecane
  • As internal standard in determination of 3-chloropropane-1,2-diol in hydrolyzed vegetable protein by capillary gas chromatography with electrolytic conductivity detection

Specifications

CAS 84539-22-0
Molecular Formula C8H10BrN3O
Linear Formula C8H10BrN3O
MDL Number MFCD00483251
Quantity 100 g
Molecular Weight (g/mol) 244.09
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.