Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Bromo-2-nitrobenzaldehyde 98.0+%, TCI America™
SDP

Catalog No. B41925G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B41925G 5 g
B41921G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B41925G Supplier TCI America Supplier No. B41925G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 20357-20-4
Molecular Formula C7H4BrNO3
Molecular Weight (g/mol) 230.017
MDL Number MFCD00456508
InChI Key UFRVBZVJVRHSNR-UHFFFAOYSA-N
PubChem CID 97233
IUPAC Name 5-bromo-2-nitrobenzaldehyde
SMILES C1=CC(=C(C=C1Br)C=O)[N+](=O)[O-]

Specifications

Melting Point 73°C
Quantity 5 g
Formula Weight 230.02
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 5-Bromo-2-nitrobenzaldehyde
RTECSNumber CU4875000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.