Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 5-Bromo-2-propoxybenzonitrile

Catalog No. OTV0001541G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
OTV0001541G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. OTV0001541G Supplier Sigma Aldrich Supplier No. OTV0001541G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

4,4,5,5,5-Pentafluoro-1-pentanol, also known as pentafluoropentanol, is a perfluorous solvent.4,4,5,5,5-Pentafluoro-1-pentanol may be used in the preparation of chlorine-capped telechelic poly(methyl methacrylate)s.

Specifications

CAS 279262-21-4
Molecular Formula C10H10BrNO
Linear Formula C10H10BrNO
MDL Number MFCD02257401
Quantity 25 g
Molecular Weight (g/mol) 240.1
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.