Learn More
Sigma Aldrich 5-bromo-2-(pyrrolidin-1-yl)pyridine
Shop All MilliporeSigma Sigma Organic Chemistry Products
Description
- 2-(3-chloropropyl)-1,3-dioxolane (2-(3α-chloropropyl)-1,3-dioxolane) is a masked α-chlorobutyraldehyde and was used for the introduction of 3-(1,3-dioxolan-2-yl)propyl moiety. it was also used in the synthesis of:
- (+/-)-histrionicotoxin and (+/-)-histrionicotoxin 235a using a two-directional strategy
- 4-iodobutyraldehyde, 5-iodovaleraldehyde and 5-iodo-2-petanone
- Corresponding phosphonatemasked α-chlorobutyraldehyde, useful for the introduction of the 3-(1,3-dioxolan-2-yl)propyl moiety; preparation and use of the corresponding phosphonate
Specifications
Specifications
| Molecular Formula | C9H11BrN2 |
| Linear Formula | C9H11BrN2 |
| MDL Number | MFCD07781213 |
| Quantity | 25 mL |
| Molecular Weight (g/mol) | 227.1 |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.