Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Bromo-2-thiopheneboronic Acid (contains varying amounts of Anhydride), TCI America™
SDP

Catalog No. B28621G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B28621G 1 g
B2862-5G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B28621G Supplier TCI America Supplier No. B28621G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Specifications

Chemical Name or Material 5-Bromo-2-thiopheneboronic Acid (contains varying amounts of Anhydride)
CAS 162607-17-2
Melting Point 135°C
Molecular Formula C4H4BBrO2S
Quantity 1 g
Synonym 5-bromothiophene-2-boronic acid, 5-bromothiophen-2-yl boronic acid, 5-bromo-2-thienylboronic acid, 5-bromo-2-thiopheneboronic acid, 2-bromothiophene-5-boronic acid, 5-bromothiophen-2-yl boranediol, 5-bromothiopheneboronic acid, 5-bromothien-2-ylboronic acid, 5-bromo-2-thienyl boronic acid, boronic acid, 5-bromo-2-thienyl
InChI Key USJPOBDLWVCPGG-UHFFFAOYSA-N
SMILES B(C1=CC=C(S1)Br)(O)O
IUPAC Name (5-bromothiophen-2-yl)boronic acid
Molecular Weight (g/mol) 206.848
PubChem CID 2734319
Formula Weight 206.85
Physical Form Crystalline Powder
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.