Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 5-Bromo-2-trifluoromethylpyridine | 436799-32-5 | MFCD06657686 | 99.7% | C6H3BrF3N | 25 G
SDP

Catalog No. 5000358699
Encompass_Preferred

5-Bromo-2-trifluoromethylpyridine is a biochemical reagent primarily for life science research. It functions as a biological material or organic compound, supporting various research applications. This product is strictly for research use and not intended for patient use.

  • Biochemical reagent for life science research
  • Can be used as a biological material or organic compound
  • Molecular weight: 225.99
  • Appearance: Solid
  • Color: White to off-white
  • Purity: 99.7%
  • For research use only

Catalog No. 50-003-58699 Supplier Medchemexpress LLC Supplier No. 25GHY66030
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

5-Bromo-2-trifluoromethylpyridine is a biochemical reagent primarily for life science research. It functions as a biological material or organic compound, supporting various research applications. This product is strictly for research use and not intended for patient use.

  • Biochemical reagent for life science research
  • Can be used as a biological material or organic compound
  • Molecular weight: 225.99
  • Appearance: Solid
  • Color: White to off-white
  • Purity: 99.7%
  • For research use only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.