Learn More
Bioworld 5-Bromo-3-Indolyl-A-D-Galactopyranoside (Blue-Gal), 100 mg
Supplier:  Bioworld 215300913
Encompass_Preferred
Chromogenic substrate for ß-D-Galactosidase. Produces a deeper blue color than X-Gal.Technical InformationMF: C14H16BrNO6MW: 374.2Cas No.: 97753-82-7Purity: >98%Melting Point: 210-213°C Soluble in DMSO and DMFProtect from light and moisture
Catalog No. 50-198-914