Learn More
Cayman Chemical 5-bromo-3-IndolylD-Galacto 1g
SDP
Catalog No. 502913548
Encompass_Preferred
A chromogenic substrate for β-galactosidase; upon enzymatic hydrolysis, it generates an indigo-colored precipitate that can be used to measure β-galactosidase activity; has been used to detect β-galactosidase in muscle from E. coli lacZ-expressing transgenic mice
Catalog No. 50-291-3548 Supplier Cayman Chemical Supplier No. 352991 G
A chromogenic substrate for β-galactosidase; upon enzymatic hydrolysis, it generates an indigo-colored precipitate that can be used to measure β-galactosidase activity; has been used to detect β-galactosidase in muscle from E. coli lacZ-expressing transgenic mice