Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 5-bromo-3-iodo-4-azaindole | 1190319-16-4 | 99.4% | 322.93 g/mol | C7H4BrIN2 | 1 G
SDP

Catalog No. 5000208593
Encompass_Preferred

5-Bromo-3-iodo-4-azaindole is an organic intermediate used in medicinal chemistry as a building block for the synthesis of biologically active compounds. It is supplied as a solid reagent for laboratory research use.

  • Molecular formula: C7H4BrIN2.
  • Molecular weight: 322.93 g/mol.
  • Cas number: 1190319-16-4.
  • Purity: 99.4%.
  • Form: solid.
  • Storage: powder at 4°C, protect from light; in solvent -80°C (6 months) or -20°C (1 month), protect from light.
  • Intended for research use only; not for human or clinical use.

Catalog No. 50-002-08593 Supplier Medchemexpress LLC Supplier No. HYW0908051G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

5-Bromo-3-iodo-4-azaindole is an organic intermediate used in medicinal chemistry as a building block for the synthesis of biologically active compounds. It is supplied as a solid reagent for laboratory research use.

  • Molecular formula: C7H4BrIN2.
  • Molecular weight: 322.93 g/mol.
  • Cas number: 1190319-16-4.
  • Purity: 99.4%.
  • Form: solid.
  • Storage: powder at 4°C, protect from light; in solvent -80°C (6 months) or -20°C (1 month), protect from light.
  • Intended for research use only; not for human or clinical use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.