Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Bromo-3-methyl-benzene-1,2-diamine, 97%, Thermo Scientific™

Catalog No. MO07383DE
Change view
Click to view available options
Quantity:
5 g
Packaging:
Amber Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
MO07383DE 5 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MO07383DE Supplier Thermo Scientific Chemicals Supplier No. MO07383DE
Add to Cart

Chemical Identifiers

CAS 76153-06-5
Molecular Formula C7H9BrN2
Molecular Weight (g/mol) 201.067
InChI Key UOFSLKHZOPVGHG-UHFFFAOYSA-N
Synonym 5-bromo-3-methyl-benzene-1,2-diamine, 5-bromo-3-methyl-1,2-benzenediamine, 1,2-benzenediamine, 5-bromo-3-methyl, 2,3-diamino-5-bromotoluene, 5-bromo-2,3-diaminotoluene, bromomethylbenzenediamine, pubchem22667, ksc642k2n, 5-bromo-2,3-diamino-toluene, 5-bromo-2.3-diamino-toluene
PubChem CID 1516397
IUPAC Name 5-bromo-3-methylbenzene-1,2-diamine
SMILES CC1=CC(=CC(=C1N)N)Br

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 5 g
Formula Weight 201.07
Percent Purity 97%
Chemical Name or Material 5-Bromo-3-methylbenzene-1, 2-diamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.