Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Bromo-3-methylpyridin-2-amine, 97%, Thermo Scientific™

Catalog No. XBX00112EA
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
XBX00112EA 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. XBX00112EA Supplier Thermo Scientific Chemicals Supplier No. XBX00112EA
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 3430-21-5
Molecular Formula C6H7BrN2
Molecular Weight (g/mol) 187.04
InChI Key KBLGGRWUEVCNPY-UHFFFAOYSA-N
Synonym 2-amino-5-bromo-3-methylpyridine, 2-amino-5-bromo-3-picoline, 2-amino-3-methyl-5-bromopyridine, 5-bromo-3-methyl-pyridin-2-ylamine, 2-pyridinamine, 5-bromo-3-methyl, 5-bromo-3-methyl-2-pyridinamine, 2-amina-5-bromo-3-picoline, 5-bromo-3-methyl-2-pyridylamine, 5-bromo-3-methyl-pyridin-2-amine, 5-bromo-3-methylpyridin-2-ylamine
PubChem CID 137934
IUPAC Name 5-bromo-3-methylpyridin-2-amine
SMILES CC1=CC(=CN=C1N)Br

Specifications

Assay Percent Range 97%
Quantity 10 g
Formula Weight 187.04
Percent Purity 97%
Chemical Name or Material 5-Bromo-3-methylpyridin-2-amine
Hazard Category Warning
Hazard Statement GHS P Statement Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.
Precautionary Statement GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.