Learn More
Medchemexpress LLC 5-Bromo-3-methylpyridin-2-ol | 89488-30-2 | 99.7% | C6H6BrNO | 10 G

Supplier: Medchemexpress LLC HY7542310G
5-Bromo-3-methylpyridin-2(1H)-one is a biochemical reagent intended for life science research. This compound appears as a white to off-white solid with a molecular weight of 188.02 and a chemical formula of C6H6BrNO.
- High purity for research applications
- Biochemical reagent for life science research
- White to off-white solid appearance
- Stable as powder at -20°C for 3 years or 4°C for 2 years
- Stable in solvent at -80°C for 6 months or -20°C for 1 month
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.