Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 5-bromo-3-phenyl salicylic acid | 99514-99-5 | 98.0% | 293.11 g/mol | C13H9BrO3 | 5 MG
SDP

Catalog No. 5000228603
Encompass_Preferred

5-Bromo-3-phenyl salicylic acid is a salicylic acid derivative for research use that acts as a selective inhibitor of human 20α-hydroxysteroid dehydrogenase (AKR1C1), with a reported Ki of 140 nM. The compound is supplied as a solid (off-white to light yellow) with a reported purity of about 98% and includes downloadable product documents (COA, SDS).

  • Selective AKR1C1 inhibitor with Ki = 140 nM.
  • Reported purity approximately 98.0%.
  • Off-white to light yellow solid suitable for laboratory handling.
  • Soluble in common organic solvents such as DMSO, ethanol, and DMF.
  • Recommended storage: -20°C sealed; in solution -80°C up to 6 months or -20°C up to 1 month.

Catalog No. 50-002-28603 Supplier Medchemexpress LLC Supplier No. HY1188195MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

5-Bromo-3-phenyl salicylic acid is a salicylic acid derivative for research use that acts as a selective inhibitor of human 20α-hydroxysteroid dehydrogenase (AKR1C1), with a reported Ki of 140 nM. The compound is supplied as a solid (off-white to light yellow) with a reported purity of about 98% and includes downloadable product documents (COA, SDS).

  • Selective AKR1C1 inhibitor with Ki = 140 nM.
  • Reported purity approximately 98.0%.
  • Off-white to light yellow solid suitable for laboratory handling.
  • Soluble in common organic solvents such as DMSO, ethanol, and DMF.
  • Recommended storage: -20°C sealed; in solution -80°C up to 6 months or -20°C up to 1 month.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.