Learn More
Medchemexpress LLC 5-bromo-3-phenyl salicylic acid | 99514-99-5 | MFCD18382111 | 98.0% | 293.11 g/mol | C13H9BrO3 | 1 MG

Supplier: Medchemexpress LLC HY1188191MG
5-Bromo-3-phenyl salicylic acid is a small-molecule research reagent that selectively inhibits human 20α-hydroxysteroid dehydrogenase (AKR1C1) with a reported Ki of 140 nM. Supplied as a solid for laboratory research, the compound should be stored sealed and protected from moisture; consult the SDS for handling and storage details.
- Selective AKR1C1 inhibitor with Ki of 140 nM.
- Additional activity: Ki 1.97 μM (AKR1C2) and 21 μM (AKR1C3).
- High purity: 98.0%.
- Molecular formula C13H9BrO3; molecular weight 293.11 g/mol.
- Appearance: off-white to light yellow solid.
- Available in small pack sizes suitable for biochemical research.
- Multilingual SDS available for safety and transport information.
- For research use only; not for human or clinical use.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.