Learn More
Medchemexpress LLC 5-Bromo-3-pyridinol | 74115-13-2 | 952150 | 100.0% | C5H4BrNO | 10 G
SDP
Catalog No. 5000433852
Encompass_Preferred
5-Bromo-3-pyridinol is a chemical and pharmaceutical intermediate used in the research of biological activities. Its chlorinated derivatives are known to be potential fungicides.
  • Chemical and pharmaceutical intermediate
  • Used in research of biological activities
  • Chlorinated derivatives are potential fungicides
  • Chlorinated derivatives provide at least 70% control of Plasmopara viticola at 200 ppm
  • Derivatives can be used to control fungal infections of plants
Catalog No. 50-004-33852 Supplier Medchemexpress LLC Supplier No. HY6605310G
5-Bromo-3-pyridinol is a chemical and pharmaceutical intermediate used in the research of biological activities. Its chlorinated derivatives are known to be potential fungicides.
  • Chemical and pharmaceutical intermediate
  • Used in research of biological activities
  • Chlorinated derivatives are potential fungicides
  • Chlorinated derivatives provide at least 70% control of Plasmopara viticola at 200 ppm
  • Derivatives can be used to control fungal infections of plants