Learn More
Bioworld X-Gluc, Sodium Salt, 500 mg
Supplier:  Bioworld 215300821
Encompass_Preferred
X-Gluc, Sodium SaltAlso known as 5-Bromo-4-Chloro-3-Indolyl-ß-D-Glucuronic Acid, Sodium Salt. Converted by the enzyme into an intense blue precipitate. It is used for the detection of the GUS gene in bacterial colonies, e.g. Escherichia Coli, or for histochemical applications.MF: C14H12BrClNO7Appearance: White, or almost white powder.Soluble in water.Protect from light and moisture.Allow to warm at room temperature before opening.Storage: dry at 2-8 C
Catalog No. 50-198-895