Learn More
Alkali Scientific 5 Bromo 4 Chloro 3 Indolyl 3 Acetate [X acetate], 200 Grams
SDP
Catalog No. 5000661226
Encompass_Preferred
Indolyl acetate is a chromogenic substrate used in enzyme detection, particularly in microbial and enzymatic assays. This 200g quantity is ideal for large-scale applications, providing a cost-effective solution for routine enzyme assays. The substrate reacts with enzymes such as esterases to produce a color change, which can be measured spectrophotometrically. It is commonly used in laboratory experiments to study enzyme activity, detect specific bacterial species, and monitor metabolic processes. Indolyl acetate is widely used in molecular biology, clinical diagnostics, and microbiology for its ability to provide sensitive and reproducible enzyme activity measurements.
Catalog No. 50-006-61226 Supplier Alkali Scientific Supplier No. OCH002CP
Indolyl acetate is a chromogenic substrate used in enzyme detection, particularly in microbial and enzymatic assays. This 200g quantity is ideal for large-scale applications, providing a cost-effective solution for routine enzyme assays. The substrate reacts with enzymes such as esterases to produce a color change, which can be measured spectrophotometrically. It is commonly used in laboratory experiments to study enzyme activity, detect specific bacterial species, and monitor metabolic processes. Indolyl acetate is widely used in molecular biology, clinical diagnostics, and microbiology for its ability to provide sensitive and reproducible enzyme activity measurements.