Learn More
Bioworld X-Gal, 1 g
Supplier:  Bioworld 215300303
Encompass_Preferred
5-Bromo-4-Chloro-3-Indolyl-β-D-Galactopyranoside A widely used histochemical substrate for beta-Galactosidase to identify lac+ bacterial colonies. Used for blue/white colony screening. Technical Information:Purity: >99% (HPLC) Soluble in DMSO and DMF Protect from light and moisture.
Catalog No. 50-198-817