Learn More
Alkali Scientific 5 Bromo 4 chloro 3 indolyl β D cellobioside [X β D Cellobioside], 1 G
SDP
Catalog No. 5000661561
Encompass_Preferred
Indolyl β-D Cellobioside (5-Bromo-4-chloro-3-indolyl β-D cellobioside) is a chromogenic substrate used in enzyme detection assays for identifying β-glucosidase and cellulase activities. This reagent allows researchers to visually track enzyme activity by producing a color change upon cleavage by the target enzyme. The 1g quantity is ideal for high-throughput screening, enzyme assays, and research into cellulose degradation. Indolyl β-D cellobioside is a valuable tool for microbiologists and biochemists studying the enzymatic breakdown of complex carbohydrates in various organisms, including bacteria, fungi, and industrial strains involved in biomass processing.
Catalog No. 50-006-61561 Supplier Alkali Scientific Supplier No. OMO003CP
Indolyl β-D Cellobioside (5-Bromo-4-chloro-3-indolyl β-D cellobioside) is a chromogenic substrate used in enzyme detection assays for identifying β-glucosidase and cellulase activities. This reagent allows researchers to visually track enzyme activity by producing a color change upon cleavage by the target enzyme. The 1g quantity is ideal for high-throughput screening, enzyme assays, and research into cellulose degradation. Indolyl β-D cellobioside is a valuable tool for microbiologists and biochemists studying the enzymatic breakdown of complex carbohydrates in various organisms, including bacteria, fungi, and industrial strains involved in biomass processing.