Learn More
Alkali Scientific 5 Bromo 4 chloro 3 indolyl β D cellobioside [X β D Cellobioside], 250 Mg
SDP
Supplier:  Alkali Scientific OMO002CP
Encompass_Preferred
Indolyl β-D Cellobioside (5-Bromo-4-chloro-3-indolyl β-D cellobioside) is a chromogenic substrate designed for enzyme detection in microbiology and biochemical research. It is commonly used to detect β-glucosidase activity, which is essential for breaking down cellulosic materials. The 250mg quantity is suitable for larger assays and can be used in screening microbial cultures or testing enzyme activity in industrial applications. This reagent produces a color change upon enzymatic cleavage, making it ideal for qualitative and quantitative enzyme assays, especially for identifying microorganisms involved in cellulose degradation or lignocellulose bioconversion processes.
Catalog No. 50-006-60003