Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Bromo-4-Chloro-3-Indolyl-β-D-Galactopyranoside , MP Biomedicals, LLC

Catalog No. MP215000150 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP215000150 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP215000150 Supplier MP Biomedicals, Inc Supplier No. 0215000150
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

5-Bromo-4-chloro-3-indolyl b-D-galactopyranoside, commonly known as X-Gal, is a histochemical substrate for b-galactosidase

  • 5-Bromo-4-chloro-3-indolyl b-D-galactopyranoside is used as indigogenic substrate for b - galactosidase, for detection of b - galactosidase-positive clones, and the identification of lac and bacterial colonies or phage plaques
  • X-Gal is a chromogenic substrate for β-galactosidase that produces a rich blue color that can easily be detected visually over background
  • X-Gal is the substrate of choice for blue-white selection of recombinant bacterial colonies with the lac+ genotype
  • X-Gal is cleaved by b-galactosidase to yield an insoluble blue precipitate
  • X-Gal is particularly useful in molecular biology applications to detect the activity of b-galactosidase which is frequently used as a reporter gene
  • Insertion of DNA into the lac Z region results in the loss of b-galactosidase activity
  • Lac+ bacterial colonies resulting from a-complementation will appear blue whereas bacterial colonies containing plasmid with DNA inserted in the lac Z region, will be incapable of a-complementation and will appear white
TRUSTED_SUSTAINABILITY

Chemical Identifiers

Molecular Weight (g/mol) 521.789

Specifications

Melting Point 230°C (decomposition)(literature)
Color White
Packaging Glass
Quantity 50 mg
Grade Not Available
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.