Learn More
Medchemexpress LLC 5-bromo-4-chloro-3-indolyl β-D-glucuronide sodium salt | 129541-41-9 | MFCD00135782 | 98.0% | 444.59 g/mol | C14H12BrClNNaO7 | 10 MG
SDP
Catalog No. 5000288669
Encompass_Preferred
X-Gluc sodium is the sodium salt of 5-bromo-4-chloro-3-indolyl β-D-glucuronide, a chromogenic substrate used to detect β-glucuronidase (GUS) activity. It reacts with GUS to produce an intense blue color and is commonly used in microbiology and molecular biology assays.
  • Chromogenic substrate for β-glucuronidase detection
  • Produces intense blue color upon enzymatic hydrolysis
  • Suitable for bacterial contamination testing and GUS reporter assays
  • High purity (98.0%) for reliable experimental results
  • Sodium salt form improves handling and aqueous solubility
Catalog No. 50-002-88669 Supplier Medchemexpress LLC Supplier No. HY15935C10MG
X-Gluc sodium is the sodium salt of 5-bromo-4-chloro-3-indolyl β-D-glucuronide, a chromogenic substrate used to detect β-glucuronidase (GUS) activity. It reacts with GUS to produce an intense blue color and is commonly used in microbiology and molecular biology assays.
  • Chromogenic substrate for β-glucuronidase detection
  • Produces intense blue color upon enzymatic hydrolysis
  • Suitable for bacterial contamination testing and GUS reporter assays
  • High purity (98.0%) for reliable experimental results
  • Sodium salt form improves handling and aqueous solubility