Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Bromo-4-chloro-3-indolyl Phosph., p-Toluidine Salt (Wht. to Off-Wht. Powd.), Fisher BioReagents

Catalog No. BP1610100 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
100 mg
500 mg
Packaging:
Amber Glass
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
BP1610-100 100 mg Amber Glass
BP1610-500 500 mg Amber Glass
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BP1610-100

missing translation for 'validMainframeMsg'

BCIP is used in conjunction with Nitro Blue Tetrazolium (NBT) in the colorimetric detection of Alkaline Phosphatase. It is suitable for use in immunohistochemistry, immunoblot staining, and ELISA applications.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

Molecular Formula C15H15BrClN2O4P
Molecular Weight (g/mol) 433.62
InChI Key QEIFSLUFHRCVQL-UHFFFAOYSA-N

Specifications

Alpha Vector Bromo-4-chloro-3-indolyl Phosphate,p-Toluidine Salt (BCIP)
CAS 6578-06-9
Melting Point 222.8°C
Color White
Assay Percent Range ≥99 %
Packaging Amber Glass
Quantity 100 mg
Synonym BCIP
Formula Weight 433.62
Percent Purity 99% min.
Physical Form Solid
Chemical Name or Material 5-Bromo-4-chloro-3-indolyl Phosphate, p-Toluidine Salt
Show More Show Less
Safety handling Check Alert storage code Red
Recommended Storage <0°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.