Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Bromo-4-Chloro-3-Indolyl Phosphate Disodium Salt, MP Biomedicals™

Catalog No. MP215004291 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
50 mg
100 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP215004291 1 g
MP215004250 50 mg
MP215004280 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. MP215004291 Supplier MP Biomedicals, Inc Supplier No. 0215004291
Add to Cart
Add to Cart

Used as a chromogenic substrate for alkaline phosphatase. 5-Bromo-4-Chloro-3-Indolyl Phosphate Disodium Salt, MP Biomedicals (BCIP) provides colormetric detection of alkaline-phosphatase-labeled molecules.

For the colorimetric detection of alkaline phosphatase-labeled molecules, 5-Bromo-4-chloro-3-indolyl phosphate (BCIP) and Nitro Blue Tetrazolium (NBT) are available.

  • The BCIP/NBT substrate system is versatile and functions in a variety of applications, including Northern Southern, and Western blotting, in situ hybridization, and immunohistochemistry
  • BCIP disodium salt is soluble in water. It may be used to prepare a stock solution, which in combination with NBT and a reaction buffer, form a substrate solution for alkaline phosphatase
  • This substrate system, when incubated with alkaline phosphatase, produces an insoluble NBT diformazan product that is easily observable with its purple color.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 102185-33-1
Molecular Formula C8H4BrClNNa2O4P
Molecular Weight (g/mol) 370.43
MDL Number MFCD00036757
InChI Key OAZUOCJOEUNDEK-UHFFFAOYSA-L
PubChem CID 6097197
IUPAC Name disodium 5-bromo-4-chloro-1H-indol-3-yl phosphate
SMILES [Na+].[Na+].[O-]P([O-])(=O)OC1=CNC2=CC=C(Br)C(Cl)=C12

Specifications

Melting Point >300°C (literature)
Quantity 1 g
Synonym sodium 5-bromo-4-chloro-1h-indol-3-yl phosphate, 5-bromo-4-chloro-3-indolyl phosphate disodium salt, 5-bromo-4-chloro-3-indoxyl phosphate, disodium salt, bcip™, 1h-indol-3-ol, 5-bromo-4-chloro-, 3-dihydrogen phosphate , sodium salt 1:2, disodium 5-bromo-4-chloro-1h-indol-3-yl phosphate, 1h-indol-3-ol,5-bromo-4-chloro-, 3-dihydrogen phosphate , sodium salt 1:2, 5-bromo-4-chloro-3-indolyl phosphate, 5-bromo-4-chloro-3-indolylphosphate disodium salt
Solubility Information Disodium salt is soluble in water (50mg/mL); insoluble in dimethylformamide.
Formula Weight 370.4
Percent Purity ≥99%
Physical Form Powder
Recommended Storage −20°C, desiccate, protect from light
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.