Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Bromo-4-chloro-3-indolyl Phosphate p-Toluidine Salt 98.0+%, TCI America™
SDP

Catalog No. B12391G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B12391G 1 g
B1239100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B12391G Supplier TCI America Supplier No. B12391G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 6578-06-9
Molecular Formula C15H15BrClN2O4P
Molecular Weight (g/mol) 433.62
MDL Number MFCD00040636
InChI Key QEIFSLUFHRCVQL-UHFFFAOYSA-N
Synonym X-Phosphate p-Toluidine Salt

Specifications

Quantity 1 g
UN Number 2811
Formula Weight 433.62
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 5-Bromo-4-chloro-3-indolyl Phosphate p-Toluidine Salt [for Biochemical Research]
TSCA Yes
Recommended Storage Freezer
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.