Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Bromo-4-chloroindoxyl 1,3-diacetate, 99%, Thermo Scientific™

Catalog No. AA4336709
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4336709 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AA4336709 Supplier Thermo Scientific Chemicals Supplier No. 4336709

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3034-04-6
Molecular Formula C16H14O3
Molecular Weight (g/mol) 254.29
MDL Number MFCD00040632
InChI Key YURQMHCZHLMHIB-UHFFFAOYNA-N
Synonym 6-methoxyflavanone, 6-methoxy-2-phenylchroman-4-one, 6-methoxy-2-phenyl-chroman-4-one, 6-methoxy-2-phenyl-2,3-dihydro-1-benzopyran-4-one, flavanone, 6-methoxy, acmc-1adv8, 6-methoxy-2-phenyl-2,3-dihydro-4h-chromen-4-one, 6-methoxy-2-phenyl-2,3-dihydro-4h-chromen-4-one #, 2,3-dihydro-6-methoxy-2-phenyl-4h-1-benzopyran-4-one, 4h-1-benzopyran-4-one, 2,3-dihydro-6-methoxy-2-phenyl
PubChem CID 97860
IUPAC Name 6-methoxy-2-phenyl-2,3-dihydrochromen-4-one
SMILES COC1=CC2=C(C=C1)OC(CC2=O)C3=CC=CC=C3

Specifications

Melting Point 165°C to 166°C
Quantity 10 g
Formula Weight 330.57
Percent Purity 99%
Physical Form Powder
Chemical Name or Material 5-Bromo-4-chloroindoxyl 1,3-diacetate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.