Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Bromo-4-chloroindoxyl Acetate 98.0+%, TCI America™
SDP

Catalog No. B483050MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
50 mg
200 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B483050MG 50 mg
B4830200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B483050MG Supplier TCI America Supplier No. B483050MG
Add to Cart
Add to Cart

Chemical Identifiers

CAS 3252-36-6
Molecular Formula C10H7BrClNO2
Molecular Weight (g/mol) 288.525
MDL Number MFCD00037932
InChI Key WPWLFFMSSOAORQ-UHFFFAOYSA-N
Synonym 5-Bromo-4-chloro-3-indolyl Acetate, 3-Acetoxy-4-chloro-5-bromoindole
PubChem CID 65116
IUPAC Name (5-bromo-4-chloro-1H-indol-3-yl) acetate
SMILES CC(=O)OC1=CNC2=C1C(=C(C=C2)Br)Cl

Specifications

Melting Point 107°C
Color White
Quantity 50 mg
Formula Weight 288.53
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 5-Bromo-4-chloroindoxyl Acetate
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.