Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Bromo-4-fluoro-2-methylaniline, 96%, Thermo Scientific Chemicals

Catalog No. AAH6248006
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6248006 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH6248006 Supplier Thermo Scientific Chemicals Supplier No. H6248006
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 627871-16-3 | Molecular Formula: C7H7BrFN | Molecular Weight: 204.04 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 627871-16-3
Molecular Formula C7H7BrFN
Molecular Weight (g/mol) 204.04
MDL Number MFCD05865218
InChI Key DNCLVDGUXUSPTL-UHFFFAOYSA-N
Synonym 2-amino-4-bromo-5-fluorotoluene, 5-bromo-4-fluoro-2-methylbenzenamine, benzenamine, 5-bromo-4-fluoro-2-methyl, 5-bromo-4-fluoro-o-toluidine, 5-bromo-4-fluoro-2-methyl-phenylamine, 2-amino-4-bromo-5-fluoro-toluene, 2-amino-4-bromo-5-fluorotoluene 5-bromo-4-fluoro-2-methylaniline, 5-bromo-4-fluoro-2-methyl-aniline, acmc-20agns, pubchem2934
PubChem CID 2782786
IUPAC Name 5-bromo-4-fluoro-2-methylaniline
SMILES CC1=CC(F)=C(Br)C=C1N

Specifications

Melting Point 86°C to 90°C
Quantity 5 g
Formula Weight 204.04
Percent Purity 96%
Physical Form Solid
Chemical Name or Material 5-Bromo-4-fluoro-2-methylaniline
Hazard Category H302-H315-H319-H335
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.