Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 5-bromo-4-methyl-thiazol-2-amine

Catalog No. SYX003051G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
200 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
SYX003051G 200 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. SYX003051G Supplier Sigma Aldrich Supplier No. SYX003051G
Only null left
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • Radiolysis of aqueous solution of chloromethane was performed with α-rays
  • Dechlorination of chloromethane on iron and palladium-iron bimetallic surface in aqueous systems has been reported

Specifications

CAS 3034-57-9
Molecular Formula C4H5BrN2S
Linear Formula C4H5BrN2S
MDL Number MFCD09260911
Quantity 200 mL
Molecular Weight (g/mol) 193.06
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.