Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Bromo-5-nitro-1,3-dioxane, ≥99.0% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111019735 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-9735 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-9735 Supplier MilliporeSigma Supelco Supplier No. 343911G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

5-Bromo-5-nitro-1,3-dioxane is a bromine containing preservative commonly used in cosmetic products.

Cleaning Products, Cosmetics, Food and Beverages and Personal Care
TRUSTED_SUSTAINABILITY

Specifications

CAS 30007-47-7
Physical Form Neat
Linear Formula C4H6 BrNO4
MDL Number MFCD00101855
Melting Point 58°C to 60°C
Type Analytical Standard
Quantity 1 g
Formula Weight 212.00
Percent Purity ≥99.0% (HPLC)
Grade Analytical Standard
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.