Learn More
Medchemexpress LLC 5-bromo-6-chloro-3-indolyl β-D-glucuronide cyclohexylammonium | 144110-43-0 | MFCD00153929 | 99.8% | 521.79 | C20H26BrClN2O7 | 5 MG
SDP
Catalog No. 5000221801
Encompass_Preferred
Magenta-β-D-GlcA (5-Bromo-6-chloro-3-indolyl β-D-glucuronide cyclohexylammonium) is a chromogenic biochemical reagent used as a substrate for β-glucuronidase assays. Upon enzymatic hydrolysis it produces a magenta-colored indoxyl product, enabling visual detection and histochemical staining in biochemical and microbiological applications. The compound is supplied as a high-purity solid in multiple pack sizes.
  • Chromogenic substrate for β-glucuronidase assays.
  • Produces a magenta-colored product for visual detection.
  • High purity: 99.79%.
  • Suitable for histochemical staining and enzyme activity assays.
  • Available in multiple pack sizes for small- to larger-scale work.
Catalog No. 50-002-21801 Supplier Medchemexpress LLC Supplier No. HY1378695MG
Magenta-β-D-GlcA (5-Bromo-6-chloro-3-indolyl β-D-glucuronide cyclohexylammonium) is a chromogenic biochemical reagent used as a substrate for β-glucuronidase assays. Upon enzymatic hydrolysis it produces a magenta-colored indoxyl product, enabling visual detection and histochemical staining in biochemical and microbiological applications. The compound is supplied as a high-purity solid in multiple pack sizes.
  • Chromogenic substrate for β-glucuronidase assays.
  • Produces a magenta-colored product for visual detection.
  • High purity: 99.79%.
  • Suitable for histochemical staining and enzyme activity assays.
  • Available in multiple pack sizes for small- to larger-scale work.