Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 5-Bromo-6-chloronicotinic acid | 29241-62-1 | 236.45 | 10 G
SDP

Catalog No. 5000445847
Encompass_Preferred

5-Bromo-6-chloronicotinic acid is an off-white to light yellow solid biochemical reagent. It is suitable for use as a biological material or organic compound in life science related research.

  • Used as a biochemical reagent
  • Suitable for life science related research
  • Molecular weight of 236.45
  • Purity of 97.34%
  • Stable for 3 years at -20°C as powder
  • Stable for 2 years at 4°C as powder
  • Stable for 6 months at -80°C in solvent
  • Stable for 1 month at -20°C in solvent

Catalog No. 50-004-45847 Supplier Medchemexpress LLC Supplier No. HYW00286310G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

5-Bromo-6-chloronicotinic acid is an off-white to light yellow solid biochemical reagent. It is suitable for use as a biological material or organic compound in life science related research.

  • Used as a biochemical reagent
  • Suitable for life science related research
  • Molecular weight of 236.45
  • Purity of 97.34%
  • Stable for 3 years at -20°C as powder
  • Stable for 2 years at 4°C as powder
  • Stable for 6 months at -80°C in solvent
  • Stable for 1 month at -20°C in solvent

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.