Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 5-Bromo-6-chloronicotinic acid | 29241-62-1 | 236.45 | 25 G
SDP

Catalog No. 5000449549
Encompass_Preferred

5-Bromo-6-chloronicotinic acid is a biochemical reagent that can be used as a biological material or organic compound for life science related research. It is for research use only.

  • Powder storage conditions: -20°C for 3 years; 4°C for 2 years.
  • Storage in solvent conditions: -80°C for 6 months; -20°C for 1 month.
  • Shipping conditions: Room temperature in continental US; may vary elsewhere.

Catalog No. 50-004-49549 Supplier Medchemexpress LLC Supplier No. 25GHYW002863
May include imposed supplier surcharges.
Add to Cart
Add to Cart

5-Bromo-6-chloronicotinic acid is a biochemical reagent that can be used as a biological material or organic compound for life science related research. It is for research use only.

  • Powder storage conditions: -20°C for 3 years; 4°C for 2 years.
  • Storage in solvent conditions: -80°C for 6 months; -20°C for 1 month.
  • Shipping conditions: Room temperature in continental US; may vary elsewhere.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.