Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Bromo-6-chloronicotinic Acid 98.0+%, TCI America™
SDP

Catalog No. B43555G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B43555G 5 g
B43551G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B43555G Supplier TCI America Supplier No. B43555G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 29241-62-1
Molecular Formula C6H2BrClNO2
Molecular Weight (g/mol) 235.44
MDL Number MFCD01927098
InChI Key DXEUARPQHJXMII-UHFFFAOYSA-M
Synonym 5-bromo-6-chloronicotinic acid, 5-bromo-6-chloro-nicotinic acid, 3-pyridinecarboxylic acid, 5-bromo-6-chloro, 3-bromo-2-chloropyridine-5-carboxylic acid, 3-bromo-2-chloro-5-pyridinecarboxylic acid, zlchem 1120, pubchem12922, acmc-1ckhn, ksc203c5j, abbypharma ap-12-5123
PubChem CID 285433
IUPAC Name 5-bromo-6-chloropyridine-3-carboxylate
SMILES [O-]C(=O)C1=CN=C(Cl)C(Br)=C1

Specifications

Melting Point 166°C
Color White-Yellow
Quantity 5 g
Formula Weight 236.45
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 5-Bromo-6-chloronicotinic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.