Learn More
Medchemexpress LLC 5-Bromo-6-chloropyrazin-2-amine | 173253-42-4 | 208.44 | 100 G
SDP
Catalog No. 5000447504
Encompass_Preferred
5-Bromo-6-chloropyrazin-2-amine (Compound 110) is an organic compound that serves as an intermediate in the synthesis of various bioactive molecules. It is promising for research related to bacterial infections, such as tuberculosis.
  • Used as an intermediate in the synthesis of bioactive molecules
  • Promising for research of bacterial infections
  • Suitable for tuberculosis research
Catalog No. 50-004-47504 Supplier Medchemexpress LLC Supplier No. HYW003686100G
Add to Cart
Add to Cart
5-Bromo-6-chloropyrazin-2-amine (Compound 110) is an organic compound that serves as an intermediate in the synthesis of various bioactive molecules. It is promising for research related to bacterial infections, such as tuberculosis.
  • Used as an intermediate in the synthesis of bioactive molecules
  • Promising for research of bacterial infections
  • Suitable for tuberculosis research