Learn More
Medchemexpress LLC 5-Bromo-6-chloropyrazin-2-amine | 173253-42-4 | 208.44 | 25 G
SDP
Catalog No. 5000448280
Encompass_Preferred
5-Bromo-6-chloropyrazin-2-amine (Compound 110) is an organic compound used as an intermediate in the synthesis of various bioactive molecules. It is promising for research of bacterial infections, such as tuberculosis.
  • Organic compound
  • Used as an intermediate in the synthesis of various bioactive molecules
  • Promising for research of bacterial infections, such as tuberculosis
Catalog No. 50-004-48280 Supplier Medchemexpress LLC Supplier No. 25GHYW003686
Add to Cart
Add to Cart
5-Bromo-6-chloropyrazin-2-amine (Compound 110) is an organic compound used as an intermediate in the synthesis of various bioactive molecules. It is promising for research of bacterial infections, such as tuberculosis.
  • Organic compound
  • Used as an intermediate in the synthesis of various bioactive molecules
  • Promising for research of bacterial infections, such as tuberculosis