Learn More
Medchemexpress LLC 5-Bromo-6-chloropyrazin-2-amine | 173253-42-4 | 208.44 | 250 G
SDP
Catalog No. 5000449923
Encompass_Preferred
5-Bromo-6-chloropyrazin-2-amine is an organic compound that serves as an intermediate in the synthesis of various bioactive molecules. This compound shows promise in the research of bacterial infections, including tuberculosis.
  • Used as an intermediate in synthesis
  • Promising for research of bacterial infections
  • High purity (99.79%)
  • Solid appearance
  • Light yellow to yellow color
Catalog No. 50-004-49923 Supplier Medchemexpress LLC Supplier No. 250GHYW003686
Add to Cart
Add to Cart
5-Bromo-6-chloropyrazin-2-amine is an organic compound that serves as an intermediate in the synthesis of various bioactive molecules. This compound shows promise in the research of bacterial infections, including tuberculosis.
  • Used as an intermediate in synthesis
  • Promising for research of bacterial infections
  • High purity (99.79%)
  • Solid appearance
  • Light yellow to yellow color