Learn More
Medchemexpress LLC 5-Bromo-7-azaindole | 183208-35-7 | 99.96% | 197.03 | 25 G
SDP
Catalog No. 5000403298
Encompass_Preferred
5-Bromo-7-azaindole is an important intermediate for the synthesis of nitrogen-containing heterocyclic compounds. This product is intended for laboratory research use only.
  • Important intermediate for synthesizing nitrogen-containing heterocyclic compounds.
  • Provided as a solid.
  • Appearance is off-white to light yellow.
  • Melting/freezing point is 179-180°C.
  • Store powder at -20°C for 3 years or 4°C for 2 years.
  • Store in solvent at -80°C for 2 years or -20°C for 1 year.
Catalog No. 50-004-03298 Supplier Medchemexpress LLC Supplier No. 25GHY30012
5-Bromo-7-azaindole is an important intermediate for the synthesis of nitrogen-containing heterocyclic compounds. This product is intended for laboratory research use only.
  • Important intermediate for synthesizing nitrogen-containing heterocyclic compounds.
  • Provided as a solid.
  • Appearance is off-white to light yellow.
  • Melting/freezing point is 179-180°C.
  • Store powder at -20°C for 3 years or 4°C for 2 years.
  • Store in solvent at -80°C for 2 years or -20°C for 1 year.