Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Bromo-7-nitroindole, 99%, Thermo Scientific™

Catalog No. AAL14923MD
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL14923MD 250 mg
AAL1492303 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL14923MD Supplier Thermo Scientific Chemicals Supplier No. L14923MD

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
5-Bromo-7-nitroindole is used in gas chromatography analysis. It is also used as pharmaceutical and medical intermediate.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 165669-16-9
Molecular Formula C8H5BrN2O2
Molecular Weight (g/mol) 241.04
MDL Number MFCD00619260
InChI Key WERRDKWLHFDNTO-UHFFFAOYSA-N
Synonym 5-bromo-7-nitroindole, acmc-20a3m9
PubChem CID 816685
IUPAC Name 5-bromo-7-nitro-1H-indole
SMILES [O-][N+](=O)C1=C2NC=CC2=CC(Br)=C1

Specifications

Melting Point 211°C to 214°C
Quantity 250 mg
Beilstein 8139934
Solubility Information Insoluble in water.
Formula Weight 241.05
Percent Purity 99%
Chemical Name or Material 5-Bromo-7-nitroindole
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.