Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Bromo-8-nitroisoquinoline 98.0+%, TCI America™
SDP

Catalog No. B41631G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B41631G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B41631G Supplier TCI America Supplier No. B41631G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 63927-23-1
Molecular Formula C9H5BrN2O2
Molecular Weight (g/mol) 253.06
MDL Number MFCD02091227
InChI Key ULGOLOXWHJEZNZ-UHFFFAOYSA-N
PubChem CID 816983
IUPAC Name 5-bromo-8-nitroisoquinoline
SMILES [O-][N+](=O)C1=C2C=NC=CC2=C(Br)C=C1

Specifications

Melting Point 140°C
Quantity 1 g
Formula Weight 253.06
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 5-Bromo-8-nitroisoquinoline
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.