Learn More
Medchemexpress LLC 5-bromo-N-[5-(4-methylphenyl)-1,3,4-oxadiazol-2-yl]thiophene-2-carboxamide | 952849-76-2 | 98.0% | 364.22 g/mol | C14H10BrN3O2S | 100 MG
SDP
Supplier:  Medchemexpress LLC HY101865100MG
Encompass_Preferred
KKL-10 is a small-molecule ribosome rescue inhibitor with broad-spectrum antimicrobial activity against bacteria. It is supplied as a research-grade solid for in vitro and mechanism-of-action studies targeting bacterial ribosome rescue pathways. Typical analytical data: CAS 952849-76-2; molecular weight 364.22 g/mol; chemical formula C14H10BrN3O2S; purity 98.0%.
  • Inhibits bacterial ribosome rescue, useful for mechanism-of-action studies.
  • Demonstrates broad-spectrum antibacterial activity against multiple strains.
  • High purity suitable for biochemical assays and screening (98.0%).
  • Provided as a solid powder with common solubility in DMSO for assay preparation.
  • Offered in multiple package sizes to support small- and mid-scale experiments.
  • For research use only; not intended for human or clinical use.
Catalog No. 50-002-45427