Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Bromo-N-methylpyridine-2-carboxamide, 96%, Thermo Scientific Chemicals

Catalog No. p-7036627
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5494706 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH5494706 Supplier Thermo Scientific Chemicals Supplier No. H5494706
Only null left
Add to Cart

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 845305-87-5
Molecular Formula C7H7BrN2O
Molecular Weight (g/mol) 215.05
MDL Number MFCD07357421
InChI Key OHYCXPXWHPWYFX-UHFFFAOYSA-N
Synonym 5-bromo-n-methylpicolinamide, n-methyl 5-bromopicolinamide, 2-pyridinecarboxamide, 5-bromo-n-methyl, n-methyl5-bromopicolinamide, acmc-209pv5, 5-bromo-n-methylpicolimide, n-methyl-5-bromopicolinamide, 5-bromo-n-methyl-2-pyridinecarboxamide
PubChem CID 22831620
IUPAC Name 5-bromo-N-methylpyridine-2-carboxamide
SMILES CNC(=O)C1=NC=C(C=C1)Br

Specifications

Melting Point 76°C to 77°C
Quantity 5 g
Formula Weight 215.05
Percent Purity 96%
Chemical Name or Material 5-Bromo-N-methylpyridine-2-carboxamide
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Keep cold; Store under Nitrogen

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.