Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 5-bromo-uridine O,S-dihydrogen phosphorothioate | 98.7% | 403.14 g/mol | C9H12BrN2O7PS | 10 MG
SDP

Catalog No. 5000298560
Encompass_Preferred

O-(((2R,3S,5R)-5-(5-bromo-2,4-dioxo-3,4-dihydropyrimidin-1(2H)-yl)-3-hydroxytetrahydrofuran-2-yl)methyl) O,S-dihydrogen phosphorothioate is a brominated nucleoside phosphorothioate provided for research and analytical applications. It has a molecular weight of 403.14 g/mol and a reported purity of 98.7%. Suitable for use in biochemical assays and synthesis workflows where modified nucleoside analogues are required.

  • Brominated nucleoside phosphorothioate structure.
  • Molecular weight 403.14 g/mol.
  • Chemical formula C9H12BrN2O7PS.
  • Purity 98.7% (as reported by distributor).
  • Available in small pack sizes for research (5 mg, 10 mg).
  • Intended for research and analytical use only.

Catalog No. 50-002-98560 Supplier Medchemexpress LLC Supplier No. HY40238310MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

O-(((2R,3S,5R)-5-(5-bromo-2,4-dioxo-3,4-dihydropyrimidin-1(2H)-yl)-3-hydroxytetrahydrofuran-2-yl)methyl) O,S-dihydrogen phosphorothioate is a brominated nucleoside phosphorothioate provided for research and analytical applications. It has a molecular weight of 403.14 g/mol and a reported purity of 98.7%. Suitable for use in biochemical assays and synthesis workflows where modified nucleoside analogues are required.

  • Brominated nucleoside phosphorothioate structure.
  • Molecular weight 403.14 g/mol.
  • Chemical formula C9H12BrN2O7PS.
  • Purity 98.7% (as reported by distributor).
  • Available in small pack sizes for research (5 mg, 10 mg).
  • Intended for research and analytical use only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.