Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Bromobenzo[b]thiophene-3-carbonyl chloride, ≥97%, Thermo Scientific™

Catalog No. CC33502DA
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CC33502DA 1 g
CC33502DE 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. CC33502DA Supplier Thermo Scientific Chemicals Supplier No. CC33502DA

Chemical Identifiers

CAS 850374-99-1
Molecular Formula C9H4BrClOS
Molecular Weight (g/mol) 275.54
MDL Number MFCD07368522
InChI Key DNAFRKYZNZZCCO-UHFFFAOYSA-N
Synonym 5-bromobenzo b thiophene-3-carbonyl chloride, benzo b thiophene-3-carbonylchloride, 5-bromo
PubChem CID 4961276
IUPAC Name 5-bromo-1-benzothiophene-3-carbonyl chloride
SMILES ClC(=O)C1=CSC2=CC=C(Br)C=C12

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 1 g
Formula Weight 275.55
Percent Purity 97%
Chemical Name or Material 5-Bromobenzo[b]thiophene-3-carbonyl chloride
Hazard Statement GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Contact with water liberates toxic gas.
Reacts violently with water.
Precautionary Statement GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place. Store in a closed container.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.