Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Bromobenzo[b]thiophene-3-carboxylic acid, 97%, Thermo Scientific™

Catalog No. CC33501CB
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CC33501CB 250 mg
CC33501DA 1 g
CC33501DE 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. CC33501CB Supplier Thermo Scientific Chemicals Supplier No. CC33501CB
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 7312-24-5
Molecular Formula C9H5BrO2S
Molecular Weight (g/mol) 257.101
InChI Key DWQBOPASRUUSKR-UHFFFAOYSA-N
Synonym 5-bromobenzo b thiophene-3-carboxylic acid, 5-bromobenzo b thiophene-3-carboxylicacid, benzo b thiophene-3-carboxylicacid, 5-bromo, benzo b thiophene-3-carboxylic acid, 5-bromo, 5-bromo-3-carboxybenzo b thiophene, 5-bromo-benzo b thiophene-3-carboxylic acid, #
PubChem CID 621037
IUPAC Name 5-bromo-1-benzothiophene-3-carboxylic acid
SMILES C1=CC2=C(C=C1Br)C(=CS2)C(=O)O

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 257.11
Percent Purity 97%
Chemical Name or Material 5-Bromobenzo[b]thiophene-3-carboxylic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.