Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Bromobenzofuran 97.0+%, TCI America™
SDP

Catalog No. B38895G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B38895G 5 g
B38891G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B38895G Supplier TCI America Supplier No. B38895G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 23145-07-5
Molecular Formula C8H5BrO
Molecular Weight (g/mol) 197.031
MDL Number MFCD03407317
InChI Key AYOVPQORFBWFNO-UHFFFAOYSA-N
Synonym 5-bromobenzofuran, benzofuran, 5-bromo, 5-bromobenzo b furan, 5-bromo-benzofuran, 5-bromobenzo beta furan, buttpark 95\50-66, pubchem22017, 5-bromobenzo b-furan, #
PubChem CID 90015
IUPAC Name 5-bromo-1-benzofuran
SMILES C1=CC2=C(C=CO2)C=C1Br

Specifications

Melting Point 8°C
Color Yellow
Boiling Point 226°C
Quantity 5 g
Formula Weight 197.03
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 5-Bromobenzofuran
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.