Learn More
Medchemexpress LLC 5-Bromoindole | 10075-50-0 | 100.0% | 196.05 | 25 G
SDP
Catalog No. 5000402954
Encompass_Preferred
5-Bromoindole is an antibacterial agent that inhibits biofilm and curli formation and reduces swarming and swimming motility. It can be used for research on enterohemorrhagic Escherichia coli O157:H7 infection.
  • Inhibits planktonic growth of Enterohemorrhagic Escherichia coli O157:H7
  • Dose-dependently inhibits biofilm formation
  • Exhibits bactericidal activity
  • Reduces swarming and swimming motility
  • Inhibits curli production
Catalog No. 50-004-02954 Supplier Medchemexpress LLC Supplier No. 25GHY30236
5-Bromoindole is an antibacterial agent that inhibits biofilm and curli formation and reduces swarming and swimming motility. It can be used for research on enterohemorrhagic Escherichia coli O157:H7 infection.
  • Inhibits planktonic growth of Enterohemorrhagic Escherichia coli O157:H7
  • Dose-dependently inhibits biofilm formation
  • Exhibits bactericidal activity
  • Reduces swarming and swimming motility
  • Inhibits curli production