Learn More
Medchemexpress LLC 5-Bromoindole | 10075-50-0 | 99.97% | 196.05 | 1 ML
SDP
Catalog No. 5000403499
Encompass_Preferred
5-Bromoindole is an antibacterial agent with a MIC of 200 μg/mL. It inhibits biofilm and curli formation, and reduces swarming and swimming motility. This compound can be used for the research of enterohemorrhagic escherichia coli o157:h7 infection.
  • Antibacterial agent
  • Inhibits biofilm and curli formation
  • Reduces swarming and swimming motility
  • Useful for enterohemorrhagic escherichia coli o157:h7 infection research
Catalog No. 50-004-03499 Supplier Medchemexpress LLC Supplier No. HY3023610MM/1ML
5-Bromoindole is an antibacterial agent with a MIC of 200 μg/mL. It inhibits biofilm and curli formation, and reduces swarming and swimming motility. This compound can be used for the research of enterohemorrhagic escherichia coli o157:h7 infection.
  • Antibacterial agent
  • Inhibits biofilm and curli formation
  • Reduces swarming and swimming motility
  • Useful for enterohemorrhagic escherichia coli o157:h7 infection research