Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 5-Bromoindole-2-carboxylic acid, 96%

Click to view available options
Quantity:
1g
5g
7254-19-5
C9H6BrNO2
240.056
MFCD00022705
YAULOOYNCJDPPU-UHFFFAOYSA-N
5-bromoindole-2-carboxylic acid,5-bromo-2-indolecarboxylic acid,1h-indolecarboxylic acid, 5-bromo,1h-indole-2-carboxylic acid, 5-bromo,pubchem1663,zlchem 1343,acmc-1bb5x,chembl22435,5-bromo indole carboxylic acid,5-bromanyl-1h-indole-2-carboxylic acid
252137
5-bromo-1H-indole-2-carboxylic acid
C1=CC2=C(C=C1Br)C=C(N2)C(=O)O
7254-19-5
C9H6BrNO2
240.056
MFCD00022705
YAULOOYNCJDPPU-UHFFFAOYSA-N
5-bromoindole-2-carboxylic acid,5-bromo-2-indolecarboxylic acid,1h-indolecarboxylic acid, 5-bromo,1h-indole-2-carboxylic acid, 5-bromo,pubchem1663,zlchem 1343,acmc-1bb5x,chembl22435,5-bromo indole carboxylic acid,5-bromanyl-1h-indole-2-carboxylic acid
252137
5-bromo-1H-indole-2-carboxylic acid
C1=CC2=C(C=C1Br)C=C(N2)C(=O)O
Quantity 5g
Beilstein 153253
Formula Weight 240.06
Physical Form Solid
Percent Purity 96%
Chemical Name or Material 5-Bromoindole-2-carboxylic acid

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

storageNote1 : -30° to -10°C