Learn More
Accela Chembio Inc 5-bromoindole-3-carboxaldehyde | 25g | 877-03-2 | MFCD00152016 | 97+% | Shelf Life: 1260 Days | Light Sensitive/store In Freezer(-20) Upon Receipt
Catalog No. 50355181
Encompass_Preferred
5-bromoindole-3-carboxaldehyde | 25g | 877-03-2 | MFCD00152016 | 97+% | Shelf Life: 1260 Days | Light Sensitive/store In Freezer(-20) Upon Receipt
Catalog No. 50-355-181 Supplier Accela Chembio Inc Supplier No. SY01446825G
5-bromoindole-3-carboxaldehyde | 25g | 877-03-2 | MFCD00152016 | 97+% | Shelf Life: 1260 Days | Light Sensitive/store In Freezer(-20) Upon Receipt